Anti-FAF1 Rabbit Polyclonal Antibody (Biotin)

BS-1348R-BIO
10299-168EA 529.29 USD
10299-168
Anti-FAF1 Rabbit Polyclonal Antibody (Biotin)
Potentiates but cannot initiate FAS-induced apoptosis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: FAF1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR