Anti-GOLPH3L Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-13489R-CY7
10299-220EA 811.1 USD
10299-220
Anti-GOLPH3L Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is localized at the Golgi stack and may have a regulatory role in Golgi trafficking.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GOLPH3L
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR