Anti-GOLGA4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-13487R-A647
10299-142EA 810.85 USD
10299-142
Anti-GOLGA4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Golgin 245 is also known as p230, GCP2, GOLG or golgi autoantigen and is a 2,230 amino acid protein that is expressed as three isoforms. Golgin 245 is localized to the cytoplasm in cells and is a member of the golgin family, all of which are peripheral membrane proteins associated with the Golgi complex. Golgin 245 has a carboxyl-terminal GRIP domain, which attaches to the trans-Golgi network (TGN) and TGN-derived vesicles. It is thought that the interaction that takes place between golgin 245 and MACF1, which cross-links microtubules to the Actin cytoskeleton, allows proteins to be transported from the TGN to the cell periphery. against golgin 245 are associated with two chronic disorders known as Sjé°ƒren's syndrome and Hepatitis B.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Golgin subfamily A member 4/Golgin 245
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR