Anti-GLG1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-13485R-FITC
10298-962EA 811.1 USD
10298-962
Anti-GLG1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Binds fibroblast growth factor and E-selectin (cell-adhesion lectin on endothelial cells mediating the binding of neutrophils).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Golgi Complex
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR