Anti-GLG1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-13485R-CY3
10298-956EA 811.1 USD
10298-956
Anti-GLG1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Binds fibroblast growth factor and E-selectin (cell-adhesion lectin on endothelial cells mediating the binding of neutrophils).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Golgi Complex
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR