Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GOLGA7 Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss
BS-13484R-BIO
GOLGA7 is a multi-pass membrane protein belonging to the Erf4 family of proteins. It is the functional ortholog of the yeast Erf4 protein. Localizing to the Golgi apparatus, GOLGA7 is a widely expressed protein but its expression is absent from colon and thymus tissues. GOLGA7 is palmitoylated on two cysteine residues, and this palmitoylation is required for its interaction with golgin 160 and its Golgi-localization. GOLGA7 also forms a complex with ZDHHC9 and, together, these proteins function as a Ras palmitoyltransferase (Ras PAT) which is required for palmitoylation of H-Ras and N-Ras proteins. The palmitoylation of Ras proteins is essential for the trafficking of Ras proteins from the Golgi to the plasma membrane, thus implicating GOLGA7 in protein transport from the Golgi to the cell surface.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: GOLGA7
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-13484R-BIO 10298-920 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol GOLGA7
Antigen Name Golgin A7
Reactivity Human, Rat, Mouse
Conjugation Biotin
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms GOLGA7A, GOLGA3AP1, HSPC041, GCP16
Gene ID 51125
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty