Anti-GOLGA6L4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-13483R-FITC
10299-086EA 810.85 USD
10299-086
Anti-GOLGA6L4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Belongs to the GOLGA6 family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GOLGA6L4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR