Anti-GOLGA6L4 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-13483R-CY3
10299-080EA 811.1 USD
10299-080
Anti-GOLGA6L4 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Belongs to the GOLGA6 family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GOLGA6L4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR