Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GNS Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-13479R-CY7
GNS is a 552 amino acid lysosomal enzyme that hydrolyzes the 6-sulfate groups of the N-acetyl-D-glucosamine 6-sulfate units of keratan sulfate and heparan sulfate. A member of the sulfatase family, GNS assists in the catabolism of heparin, and binds calcium as a cofactor. GNS deficiency results in an autosomal recessive lysosomal storage disorder known as mucopolysaccharidosis type IIID (Sanfilippo D syndrome), which is characterized by mild somatic disease and severe degeneration of the central nervous system. Subject to post-translational internal peptidase cleavage, GNS is encoded by a gene mapping to human chromosome 12q14.2 and mouse chromosome 10 D2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNS/Glucosamine 6 sulfatase
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-13479R-CY7 10298-888 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol GNS
Antigen Name Glucosamine (N-acetyl)-6-sulfatase
Reactivity Mouse, Human, Rat
Conjugation Cy7®
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms Glucosamine-6-sulfatase, G6S, GNS, N-acetylglucosamine-6-sulfatase
Gene ID 2799
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty