Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GNPDA2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
During fertilization in mammals, the sperm activates the egg by causing an increase in the level of free cytoplasmic calcium concentration. This increased calcium concentration induces a characteristic series of oscillations that trigger egg activation and early embryo development. A hamster protein named oscillin is thought to be involved in this pathway. The enzyme glucosamine-6-phosphate isomerase (GNPI) or deaminase (GNPDA1) and the related protein GNPDA2 are the human homologs of hamster oscillin. GNPDA1 and GNPDA2 catalyze the conversion of GNP to fructose-6-phosphate and ammonia. Both proteins exist as homohexamers and are ubiquitously expressed with highest expression in testis, ovary and heart. Three isoforms of GNPDA2 are expressed due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNPDA2
Clonality: Polyclonal
Conjugation: Cy5.5®
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL 13474R-CY5.5 10227-004 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol GNPDA2
Antigen Name Glucosamine-6-phosphate Deaminase 2
Reactivity Mouse, Rat, Human
Conjugation Cy5.5®
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms GNPDA 2, SB52, Glucosamine-6-phosphate isomerase 2, GNP2, Glucosamine-6-phosphate deaminase 2
Gene ID 132789
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty