Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GNL3L Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-13472R-A350
GTPases from the MMR1/HSR1 GTP-binding protein subfamily are circularly rearranged G-motifs that play a critical role in maintaining normal cell growth. Deletion of these genes results in severe growth defects with a marked reduction in mature rRNA species and a concomitant accumulation of the 35S pre-rRNA transcript. Deletion also causes the ribosomal protein Rpl25a to fail exportation from the nucleolus. Deletion of any of the G-domain motifs will result in a null phenotype and nuclear/nucleolar localization that lacks the nucleolar export of preribosomes and is accompanied by a distortion of the nucleolar structure. GNL3L (guanine nucleotide binding protein-like 3 (nucleolar)-like) is a 582 amino acid nuclear protein that belongs to the MMR1/HSR1 GTP-binding protein family. Containing one G (guanine nucleotide-binding) domain, GNL3L is required for normal processing of ribosomal pre-rRNA and for cell proliferation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNL3L
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-13472R-A350 10298-636 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol GNL3L
Antigen Name Guanine Nucleotide Binding Protein-like 3 (nucleolar)-like
Reactivity Human, Mouse, Rat
Conjugation Alexa Fluor® 350
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms novel GTPase, OTTHUMP00000023374., GNL3L_HUMAN, FLJ10613, Guanine nucleotide binding protein-like 3 nucleolar-like
Gene ID 54552
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty