Anti-GNA14 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-13464R-CY3
10298-572EA 746.19 USD
10298-572
Anti-GNA14 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
This gene encodes a member of the guanine nucleotide-binding, or G protein family. G proteins are heterotrimers consisting of alpha, beta and gamma subunits. The encoded protein is a member of the alpha family of G proteins, more specifically the alpha q subfamily of G proteins. The encoded protein may play a role in pertussis-toxin resistant activation of phospholipase C-beta and its downstream effectors.[provided by RefSeq, Feb 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNA14
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR