Anti-GMPR2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-13463R-CY5
10298-552EA 811.1 USD
10298-552
Anti-GMPR2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
GMPR2

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GMPR2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR