Anti-GMPR1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-13462R-A555
10298-522EA 811.1 USD
10298-522
Anti-GMPR1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
This gene encodes an enzyme that catalyzes the irreversible and NADPH-dependent reductive deamination of GMP to IMP. The protein also functions in the re-utilization of free intracellular bases and purine nucleosides.[provided by RefSeq, Oct 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GMPR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR