Anti-GMPPA Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-13460R-A647
10298-306EA 811.1 USD
10298-306
Anti-GMPPA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
GMPPA is a GDP-mannose pyrophosphorylase which catalyzes the conversion of mannose-1-phosphate and GTP to GDP-mannose, in the production of N-linked oligosaccharides.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GMPPA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR