Anti-OBP1F Rabbit Polyclonal Antibody (Biotin)

BS-12488R-BIO
10282-508EA 529.29 USD
10282-508
Anti-OBP1F Rabbit Polyclonal Antibody (Biotin)
This protein is found in nasal epithelium and it binds a wide variety of chemical odorants.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: OBP1F
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR