Anti-OBP1F Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-12488R-A488
10282-502EA 811.1 USD
10282-502
Anti-OBP1F Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
This protein is found in nasal epithelium and it binds a wide variety of chemical odorants.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OBP1F
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR