AGPS is a 658 amino acid enzyme that is required for glycerolipid metabolism and ether lipid biosynthesis. Localized to the inner aspect of the peroxisomal membrane, AGPS is likely part of a heterotrimeric complex that is also composed of GNPAT and a modified form of GNPAT. Containing one FAD-binding PCMH-type domain, AGPS utilizes FAD as a cofactor in the synthesis of alkyl-glycerone 3-phophate and a long-chain acid anion from 1-acteyl-glyerone 3-phosphate and a long-chain alcohol. Defects in the gene encoding AGPS results in rhizomelic chondrodysplasia punctata type 3, a disease characterized by vertebral disorders, severe mental retardation, cutaneous lesions, cataracts and rhizomelic shortening of the humerus and femur.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: AGPS
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat