Anti-ALDH9A1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-12461R-CY5
10282-192EA 811.1 USD
10282-192
Anti-ALDH9A1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Aldehyde dehydrogenases (ALDHs) mediate the NADP+-dependent oxidation of aldehydes into acids and play an important role in the detoxification of alcohol-derived acetaldehyde, as well as in lipid peroxidation and in the metabolism of corticosteroids, biogenic amines and neurotransmitters. ALDH9A1 (aldehyde dehydrogenase family 9 member A1), also known as E3, ALDH4, ALDH7, ALDH9 or TMABADH, is a 494 amino acid cytoplasmic protein that is highly expressed in adult liver, skeletal muscle, kidney and embryonic brain. ALDH9A1 converts gamma-trimethylaminobutyraldehyde into gamma-butyrobetaine and catalyzes the irreversible oxidation of a broad range of aldehydes to the corresponding acids in a NAD-dependent reaction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALDH9A1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR