LRRFIP2 is a 721 amino acid protein that belongs to the LRRFIP family. Ubiquitously expressed, LRRFIP2 acts as an activator of the Wnt signaling pathway and as a positive regulator of NFâ…¹B activity. LRRFIP2 may be involved in regulating cytokine production in macrophages, suggesting a functional role in the TLR4-mediated inflammatory response. Three isoforms of LRRFIP2 exists due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LRRFIP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat