Anti-LRRFIP2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-12438R-CY5
10283-918EA 811.1 USD
10283-918
Anti-LRRFIP2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
LRRFIP2 is a 721 amino acid protein that belongs to the LRRFIP family. Ubiquitously expressed, LRRFIP2 acts as an activator of the Wnt signaling pathway and as a positive regulator of NFâ…¹B activity. LRRFIP2 may be involved in regulating cytokine production in macrophages, suggesting a functional role in the TLR4-mediated inflammatory response. Three isoforms of LRRFIP2 exists due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRFIP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR