Anti-FRAT2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-12435R-A647
10281-856EA 811.1 USD
10281-856
Anti-FRAT2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
FRAT1 and FRAT2 were originally characterized as proteins frequently rearranged in advanced T cell lymphoma, and they have since been identified as proto-oncogenes involved in tumorigenesis. These proteins share significant homology with the Xenopus glycogen synthase kinase-3 (xGSK-3) binding protein, which is designated GBP and is essential for the formation of the dorsal-ventral axis during embryonic development. Establishment of these embryonic axes is mediated by the Wnt intracellular signaling pathway. Wnt signaling is regulated in part by the activity of GSK-3, which phosphorylates and thereby facilitates the degradation of ?catenin. GBP binds to GSK-3 and inhibits this phosphorylation, resulting in the accumulation of ?catenin and the subsequent transcription of Wnt target genes. Like GBP, FRAT2 has been shown to bind xGSK-3, suggesting that FRAT1 and FRAT2 may be GSK-3 regulatory proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FRAT2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR