Anti-DOCK5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-12433R-A488
10281-808EA 746.42 USD
10281-808
Anti-DOCK5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
DOCK 5 is a 1,870 amino acid protein belonging to the DOCK family of cytokinesis-regulating proteins. This cytoplasmic peripheral membrane protein activates Rac 1 and Rac 2 small GTPases, while presumably acting as a guanine nucleotide exchange factor (GEF), which exchanges bound GDP for free GTP. DOCK 5 contains one DHR-1 (CZH-1) domain, one DHR-2 (CZH-2) domain and one SH3 domain. The DHR-2 domain is a putative GEF activity mediator. In mice, spontaneous mutation of the gene encoding DOCK 5 leads to deletion of the DHR-1 domain, which functions to bind phospholipids and assists in protein-protein interactions, resulting in rupture of lens cataract (RLC). Due to siRNA knockdown studies, it is suspected that DOCK 5 may also be an important mediator of CrkII/CrkL regulation of Caco-2 migration and spreading on COL4. There are two isoforms of DOCK 5 that exist as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DOCK5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR