Anti-FLVCR1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-12344R-CY5
10278-884EA 776.17 USD
10278-884
Anti-FLVCR1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
FLVCR is responsible for the exportation of cytoplasmic heme groups. It is believed that it may protect developing erythroid cells from heme toxicity. Expression of FLVCR in cells will cause susceptibility to FeLV-C (Feline leukemia virus subgroup C) in vitro. FLVCR is found in all hematopoietic tissues, including peripheral blood lymphocytes and fetal liver, and some expression is found in pancreas and kidney. It is down-regulated in haemopoietic progenitor cells undergoing differentiation and hemoglobinization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FLVCR
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR