Anti-DNER Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-12115R-CY3
10272-808EA 811.1 USD
10272-808
Anti-DNER Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Activator of the NOTCH1 pathway. May mediate neuron-glia interaction during astrocytogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DNER
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR