Anti-NFIX Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-11900R-CY5
10267-596EA 811.1 USD
10267-596
Anti-NFIX Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Recognizes and binds the palindromic sequence 5'-TTGGCNNNNNGCCAA-3' present in viral and cellular promoters and in the origin of replication of adenovirus type 2. These proteins are individually capable of activating transcription and replication.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NFIX
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR