Anti-GAST Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-1189R-A350
10267-560EA 811.1 USD
10267-560
Anti-GAST Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Gastrin stimulates the stomach mucosa to produce and secrete hydrochloric acid and the pancreas to secrete its digestive enzymes. It also stimulates smooth muscle contraction and increases blood circulation and water secretion in the stomach and intestine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Gastrin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR