Anti-NAV2/RAINB1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-11892R-CY5
10267-406EA 811.1 USD
10267-406
Anti-NAV2/RAINB1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Neuron navigator 2 (NAV2), also known as RAINB1 (retinoic acid inducible in neuroblastoma 1), POMFIL2 (pore membrane and/or filament-interacting-like protein 2), HELAD1 (helicase APC down-regulated 1), unc53H2 or steerin-2, is a 2,488 amino acid nuclear protein that plays a role in the development of sensory organs. Existing as thirteen alternatively spliced isoforms, Neuron navigator 2 displays 3' to 5' helicase activity and exonuclease activity and participates in the catalysis of ATP to ADP. Neuron navigator 2 is highly expressed in the nervous system of developing embryos and colon carcinomas, as well as in liver, brain and kidney. Lower levels of expression are found in lung, thyroid, spinal cord, heart, placenta and mammary gland. Neuron navigator 2 belongs to the Nav/unc-53 family and contains one CH (calponin-homology) domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NAV2/RAINB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR