Anti-MPP5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-11887R-A555
10267-336EA 811.1 USD
10267-336
Anti-MPP5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Two highly conserved complexes are responsible for the assembly of tight junctions, the Crumbs-Pals1-Patj complex and the Cdc42-Par6-Par3-aPKC complex. Tight junctions assist in the formation of polarity in the epithelia by establishing a barrier to separate apical and basolateral membranes. Pals1, importantly, mediates interaction between the two complexes via interaction with Par6. Loss of Pals1 function results in delayed polarization, decreased transepithelial electrical resistance and an inability to form lumenal cysts. Because tumors exhibit perturbations in epithelial polarity, Pals1 presents a new potential target in the study of carcinogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MPP5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR