IL-1R9 is a member of the interleukin-1/Toll-like receptor family expressed in fetal brain. It is a single pass transmembrane protein with a highly conserved cytoplasmic region that is slightly longer than that of other family members. IL-1R9 is highly homologous to IL-1R8 and the IL-1 accessory protein receptor chains. The genes encoding IL-1R9 and IL-1R8 are both found on the X-chromosome in the region associated with X-linked non-syndromic mental retardation. Unlike other family members, IL-1R9 and IL-1R8 do not activate NF-kB.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL1RAPL2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat