Anti-HMX2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-11853R-A488
10264-746EA 811.1 USD
10264-746
Anti-HMX2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
The homeobox DNA-binding domain is a 60 amino acid motif that is conserved among many species and functions to bind DNA via a helix-turn-helix structure, thereby playing a role in transcriptional regulation and the control of gene expression. HMX2 (H6 family homeobox 2), also known as H6L or Nkx5-2, is a 273 amino acid nuclear protein that belongs to the HMX homeobox family and contains one homeobox DNA-binding domain. HMX2 functions as a transcription factor that assists in specification of neuronal cell types and is essential for proper development of hypothalamus and inner ear. Hemizygous deletions of the gene encoding HMX2 are thought to lead to vestibular dysfunction, inner ear malformations and congenital sensorineural hearing loss.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HMX2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR