Anti-GBX2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-11849R-A488
10264-634EA 810.85 USD
10264-634
Anti-GBX2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
The isthmic organizer signals at the mid/hindbrain boundary (MHB) regulate the development and differentiation of the vertebrate caudal midbrain and the anterior hindbrain. The MHB forms at the boundary of expression between homeobox genes Gbx2 and Otx2. Gbx2 and Otx2 play distinct, essential roles in MHB positioning and development. During development, the GBX2 gene is expressed in the anterior hindbrain. Specifically, Gbx2 negatively regulates Otx2 expression along the anterior-posterior axis; Gbx2(-) mutants demonstrate an expanded Otx2 domain. During development, the GBX2 gene is expressed in the anterior hindbrain. Gbx2 is expressed in the adult brain, spleen and female genital tract. The GBX2 gene is over-expressed in human prostate cancer cell lines (TSU-prl, PC3, DU145 and LNCaP). Furthermore, downregulation of Gbx2 expression restricts tumorigenicity in human prostate cancer cell lines, which suggests that Gbx2 expression may be required for growth of malignant prostate cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Gbx2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR