Anti-FNBP1L Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-11841R-CY3
10264-510EA 811.1 USD
10264-510
Anti-FNBP1L Rabbit Polyclonal Antibody (Cy3®)
Antibodies
TOCA-1 is a 605 amino acid protein that localizes to the cytoplasm and the cytoskeleton, as well as to cytoplasmic vesicles and the cell membrane, and contains one FCH domain, one REM repeat and one SH3 domain. Existing as multiple alternatively spliced isoforms, TOCA-1 interacts with CDC42 and is required for the coordination of membrane tubulation with Actin cytoskeletal reorganization during endocytosis. Additionally, TOCA-1 is involved in membrane invagination, tubule formation and Actin polymerization. The gene encoding TOCA-1 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FNBP1L/TOCA1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR