Anti-IL17A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-1183R-A488
10267-100EA 811.1 USD
10267-100
Anti-IL17A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Induces stromal cells to produce proinflammatory and hematopoietic cytokines. Enhances the surface expression of ICAM1/intracellular adhesion molecule 1 in fibroblasts.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL17A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR