Anti-DNTNP/FAM53A Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-11839R-A350
10264-466EA 811.1 USD
10264-466
Anti-DNTNP/FAM53A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Dorsal neural-tube nuclear protein is a 398 amino acid nuclear protein. DNTNP is thought to play an important role in neural development by specifying dorsal cell fates within the neural tube. Widely expressed in the dorsal neural tube, DNTNP is most highly expressed in the dorsal regions of the midbrain, the hindbrain, the diencephalon, and the spinal neural tube and is expressed at lower levels in the branchial arches, the telencephalon, the heart, and somites of developing embryos. DNTNP is encoded by a gene located on chromosome 4 which contains many genes including the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DNTNP/FAM53A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR