Anti-CUX2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-11832R-CY3
10264-332EA 811.1 USD
10264-332
Anti-CUX2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
May be a transcription factor involved in neural specification. Binds to DNA in a sequence-specific manner (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CUX2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR