Anti-DRGX Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-11827R-CY5
10264-248EA 811.1 USD
10264-248
Anti-DRGX Rabbit Polyclonal Antibody (Cy5®)
Antibodies
DRGX is a transcription factor required for the formation of correct projections from nociceptive sensory neurons to the dorsal horn of the spinal cord and normal perception of pain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DRGX
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR