Anti-DRGX Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-11827R-A555
10264-240EA 811.1 USD
10264-240
Anti-DRGX Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
DRGX is a transcription factor required for the formation of correct projections from nociceptive sensory neurons to the dorsal horn of the spinal cord and normal perception of pain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DRGX
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR