Anti-DOCK7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-11825R-A647
10264-204EA 811.1 USD
10264-204
Anti-DOCK7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
DOCK 7 is a 2,140 amino acid protein that localizes to developing axons and contains one DHR-1 domain and one DHR-2 domain. Expressed in a variety of tissues, DOCK 7 functions as a guanine nucleotide exchange factor (GEF) that specifically activates Rac 1 and Rac 3 by catalyzing the exchange of bound GDP for free GTP. Multiple isoforms of DOCK 7 exist due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DOCK7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR