Anti-CAPS1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-11812R-FITC
10263-206EA 757.82 USD
10263-206
Anti-CAPS1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Calcium-dependent secretion activators (CAPS-1 and CAPS-2) are calcium-binding proteins that direct neurotransmitter and neuropeptide-filled vesicles to the cell membrane for secretory granule exocytosis. Both CAPS-1 and CAPS-2 are expressed primarily in the brain where they regulate the secretion of various substances. The CAPS proteins contain a PH domain that is essential for regulation of exocytosis, as well as regulation of phospholipid binding. Through their regulation of neurotrophin release from granule cells, CAPS proteins help to regulate cell fate during neuronal development. CAPS-1 is thought to regulate catecholamine release from neuronal cells, while CAPS-2 is thought to regulate release of both brain-derived neurotrophic factor (BDNF) and neurotrophin-3 (NT-3) from granule cells. Defects in the genes encoding CAPS-1 and CAPS-2 are implicated in impaired cerebral development and autism.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAPS1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR