Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AIP1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-11705R-CY7
AIP1 (ASK-interacting protein 1), also known as DAB2IP (DAB2 interacting protein), is a 1,189 amino acid protein that localizes to both the membrane and the cytoplasm and contains one Ras-GAP domain, one PH domain and one C2 domain. Expressed at low levels in prostate tissue, AIP1 functions as a Ras GTPase-activating protein that interacts with ASK 1 and, via this interaction, disrupts the association of ASK 1 with an inhibitory 14-3-3 complex, thereby allowing free ASK 1 to function within the cell. AIP1 exists as multiple alternatively spliced isoforms and is down-regulated in prostate cancer tissue, suggesting a role in tumor suppression. Chromosomal aberrations in the gene encoding AIP1 are associated with acute myeloid leukemia (AML), implicating a role for AIP1 fusion proteins in tumorigenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AIP1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-11705R-CY7 10259-808 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol AIP1
Antigen Name Activin receptor interacting protein 1
Reactivity Rat, Human, Mouse
Conjugation Cy7®
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms Membrane-associated guanylate kinase inverted 2, Membrane associated guanylate kinase 2, MAGI2, Acvri1, ACVRIP1, ARIP1, Membrane associated guanylate kinase WW and PDZ domain containing protein 2, AIP 1, MAGI2_HUMAN, Atrophin 1 interacting protein 1, Atrophin-1-interacting protein 1, Atrophin-1-interacting protein A, Acvrinp1, Aip-1, Membrane associated guanylate kinase WW and PDZ domain containing 2, SSCAM, Activin receptor interacting protein 1, WW and PDZ domain-containing protein 2., Membrane-associated guanylate kinase, Membrane associated guanylate kinase inverted 2, Atrophin 1 interacting protein A, MAGI-2, S SCAM
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty