Anti-HTT Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-11699R-A555
10263-072EA 811.1 USD
10263-072
Anti-HTT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
May play a role in microtubule-mediated transport or vesicle function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HTT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR