Anti-TMEM106B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-11694R-FITC
10262-050EA 811.1 USD
10262-050
Anti-TMEM106B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Involved in dendrite morphogenesis and maintenance by regulating lysosomal trafficking via its interaction with MAP6. May act by inhibiting retrograde transport of lysosomes along dendrites. Required for dendrite branching.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMEM106B
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR