Anti-FGGY Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-11689R-CY7
10261-990EA 811.1 USD
10261-990
Anti-FGGY Rabbit Polyclonal Antibody (Cy7®)
Antibodies
This gene encodes a protein that phosphorylates carbohydrates such as ribulose, ribitol, and L-arabinitol. Genome-wide association studies in some populations have found an association between polymorphisms in this gene and sporadic amyotrophic lateral sclerosis, but studies of other populations have not been able to replicate this association. Alternative splicing results in multiple transcript variants. [provided by RefSeq, May 2013]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGGY
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR