Anti-THOP1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-11683R-CY7
10261-876EA 811.1 USD
10261-876
Anti-THOP1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Thimet oligopeptidase, also designated soluble metallo-endopeptidase, is a cytoplasmic protein belonging to the peptidase M3 family. The gene for the protein maps against chromosome 19q13.3. Thimet oligopeptidase can degrade the b-Amyloid precursor protein and generate amyloidogenic fragments. It is important in cytoplasmic peptide degradation and involved in metabolism of neuropeptides that are less than 20 amino acids in length. Thimet oligopeptidase is highly expressed in testis but can also be detected in liver, lung and kidney.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: THOP1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR