Anti-APBA2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-11651R-A555
10260-910EA 811.1 USD
10260-910
Anti-APBA2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
The Beta-Amyloid precursor protein (Beta-APP) is a major constituent of the amyloid deposits in patients with Alzheimer’s disease. The Beta-Amyloid precursor is known to interact with several proteins, including X11 and the G heterotrimetric protein APP-BP1. The neuronal, transmembrane protein X11 is known to bind to the ∫-Amyloid precursor protein via a phosphotyrosine binding (PTB) domain, reducing the secretion of cellular Beta-APP and slowing Beta-APP processing pathways. X11 binds specifically to the YENPTY motif, which is involved in the internalization of Beta-APP. Multiple splice varitents of X11 have been identified, including X11 Alpha (also designated Mint 1), X11 Beta (Mint 2) and X11 Gamma(Mint 3).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APBA2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR