Anti-TRAK2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-11650R-HRP
10260-902EA 811.1 USD
10260-902
Anti-TRAK2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
ALS2CR3 may regulate endosome-to-lysosome trafficking of membrane cargo, including EGFR.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: TRAK2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR