Anti-CALHM1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-11642R-A647
10260-406EA 810.85 USD
10260-406
Anti-CALHM1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
CALHM1 is a 346 amino acid multi-pass endoplasmic reticulum membrane protein that belongs to the FAM26 family. CALHM1 co-localizes with GRP 78 to the endoplasmic reticulum. Predominantly expressed in adult brain, CALHM1 may be a pore-forming ion channel that controls cytosolic Ca2+ permeability and cytosolic Ca2+ concentration in the cell. It is suggested that CALHM1 regulates amyloid precursor protein proteolysis and aggregated amyloid-beta peptides levels in a Ca2+ dependent manner. CALHM1 homomultimerizes and shares strong sequence similarities with the selectivity filter of the NMDA receptor, which generates a large Ca2+ conductance across the plasma membrane. CALHM1 may be a potential factor involved in the pathogenesis of Alzheimer's disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CALHM1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR