Anti-OR5P3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-11628R-A350
10260-202EA 811.1 USD
10260-202
Anti-OR5P3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Odorant receptor (Potential). May be involved in taste perception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OR5P3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR