Anti-PIRT Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-11623R-CY3
10263-460EA 811.1 USD
10263-460
Anti-PIRT Rabbit Polyclonal Antibody (Cy3®)
Antibodies
PIRT is a 137 amino acid multi-pass membrane protein. Highly conserved among vertebrates, PIRT consists of two transmembrane domains and one putative C-terminal phosphoinositide-binding domain. Although PIRT is expressed in peripheral nervous system, with highest levels in dorsal root ganglion and trigeminal neurons, and lowest levels in sympathetic and enteric neurons, it is not expressed in spinal cord. PIRT is a required component of the VR1 complex, which positively regulates VR1, a sensor of both noxious heat and capsaicin. Correspondingly, PIRT knockout results in impaired responses to noxious heat and capsaicin exposure, while VR1 remains unaltered. The gene that encodes PIRT maps to human chromosome 17p13.1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PIRT
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR